missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chlorocyclopentane, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2502336

 View more versions of this product

Catalog No. AAB2502336


Explore available promotions

Description

Description

Chlorocyclopentane was used in the preparation of diastereomeric activation products. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorocyclopentane was used in the preparation of diastereomeric activation products. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in water. 760 mg/L.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
Specifications

Specifications

Chlorocyclopentane
-94°C
113°C to 114°C
C5H9Cl
MFCD00001360
UN1993
cyclopentane, chloro, cyclopentyl chloride, chloroclopentane, cyclopentylchloride, unii-to3f41cl9a, 1-chlorocyclopentane, to3f41cl9a, cyclopentylchlorid, chloro-cyclopentan, chlorocyclopenetane
NDTCXABJQNJPCF-UHFFFAOYSA-N
chlorocyclopentane
70252
99%
930-28-9
1.003
15°C (59°F)
1.4512
500 g
1900557
Soluble in water. 760mg/L.
C1CCC(C1)Cl
104.577
104.58

Chemical Identifiers

930-28-9
104.577
NDTCXABJQNJPCF-UHFFFAOYSA-N
70252
C1CCC(C1)Cl
C5H9Cl
MFCD00001360
cyclopentane, chloro, cyclopentyl chloride, chloroclopentane, cyclopentylchloride, unii-to3f41cl9a, 1-chlorocyclopentane, to3f41cl9a, cyclopentylchlorid, chloro-cyclopentan, chlorocyclopenetane
chlorocyclopentane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225
Highly flammable liquid and vapour.

P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P332+P313-P363-P370+P378q-P501c

H225-H315-H319

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 213-212-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only