Learn More
Chlorodimethyl-n-octadecylsilane, 95%, Thermo Scientific Chemicals
Preparation of hydrophobic surfaces, chain terminator
Supplier: Thermo Scientific Chemicals 04241422
Description
Chlorodimethyl-n-octadecylsilane is used for preparation of hydrophobic surfaces and chain terminator.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
184°C to 186°C (0.2mmHg) | |
28°C to 32°C | |
White | |
100 g | |
MFCD00000504 | |
2246012 | |
chlorodimethyloctadecylsilane, dimethyloctadecylchlorosilane, chlorodimethyl octadecyl silane, octadecyldimethylchlorosilane, silane, chlorodimethyloctadecyl, n-octadecyldimethylchlorosilane, chloro dimethyl octadecylsilane, chlorodimethyl-n-octadecylsilane, dimethyl-n-octadecylchlorosilane, stearyldimethylchlorosilane | |
GZGREZWGCWVAEE-UHFFFAOYSA-N | |
chloro-dimethyl-octadecylsilane | |
87738 | |
95% | |
1.453 | |
0.850 g/mL |
Chlorodimethyl-n-octadecylsilane | |
18643-08-8 | |
Solid | |
C20H43ClSi | |
UN3261 | |
200°C (392°F) | |
Reacts with water. | |
CCCCCCCCCCCCCCCCCC[Si](C)(C)Cl | |
347.10 | |
347.1 | |
Acrid | |
Moisture sensitive |
Chemical Identifiers
18643-08-8 | |
347.10 | |
GZGREZWGCWVAEE-UHFFFAOYSA-N | |
87738 | |
CCCCCCCCCCCCCCCCCC[Si](C)(C)Cl |
C20H43ClSi | |
MFCD00000504 | |
chlorodimethyloctadecylsilane, dimethyloctadecylchlorosilane, chlorodimethyl octadecyl silane, octadecyldimethylchlorosilane, silane, chlorodimethyloctadecyl, n-octadecyldimethylchlorosilane, chloro dimethyl octadecylsilane, chlorodimethyl-n-octadecylsilane, dimethyl-n-octadecylchlorosilane, stearyldimethylchlorosilane | |
chloro-dimethyl-octadecylsilane |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 242-472-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only