Learn More
Chloroform, 99.9%, Extra Dry, stabilized, AcroSeal™, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 326820010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Chloroform | |
Water <50 ppm, Stabilized | |
1.4800g/mL | |
1 L | |
Liquid | |
99.9% | |
CHCl3 | |
MFCD00000826 | |
1888 | |
01, 61 | |
15, 2142 | |
Solubility in water: 8g/L (20°C). Other solubilities: miscible with alcohol, benzene, ether, petr.ether,, carbon tetrachloride, carbon disulfide and oils., soluble in acetone. | |
ClC(Cl)Cl | |
119.37 | |
CHEBI:35255 | |
99.9% |
-63.0°C | |
67-66-3,513-35-9 | |
61.0°C | |
AcroSeal™ Glass Bottle | |
0.0005% max. | |
1.48 | |
CHCl3 | |
0.56 mPa.s (20°C) | |
0.005% max. (Coulometric) | |
01,130; 08,92; 15,84 | |
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio | |
HEDRZPFGACZZDS-UHFFFAOYSA-N | |
chloroform | |
6212 | |
119.38 | |
Extra Dry |
Chemical Identifiers
67-66-3 | |
119.37 | |
HEDRZPFGACZZDS-UHFFFAOYSA-N | |
6212 | |
ClC(Cl)Cl |
CHCl3 | |
MFCD00000826 | |
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio | |
CHEBI:35255 |
Safety and Handling
GHS H Statement
Causes skin irritation. Causes serious eye irritation. Harmful if swallowed. Toxic if inhaled. Suspected of causing cancer. May cause drowsiness or dizziness. Suspected of damaging the unborn child. Causes damage to organs through prolonged or repeated exposure if inhaled.
GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician. IF ON SKIN: Wash with plenty of soap and water. Do not breathe dust/fume/gas/mist/vapors/spray. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection.
Danger
RUO – Research Use Only