missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chloroform, 99+%, for spectroscopy, stabilized with amylene, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 167735000

 View more versions of this product

Catalog No. AC167735000



Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For UV/VIS spectroscopy
Specifications

Specifications

Chloroform
Stabilized with amylene
0.005 max. at 280nm, 0.01 max. at 270nm, 0.075 max. at 260nm, 0.5 max. at 250nm, 1 max. at 245nm
61.0°C
Glass Bottle
0.0003% max.
1.48
CHCl3
0.56 mPa.s (20°C)
01, 61
15, 2142
Solubility in water: 8g/L (20°C). Other solubilities: miscible with alcohol, benzene, ether, petr.ether,, carbon tetrachloride, carbon disulfide and oils., soluble in acetone.
ClC(Cl)Cl
119.37
CHEBI:35255
≥99%
-63.0°C
67-66-3,513-35-9
1.4800g/mL
500 mL
Liquid
99+%
CHCl3
MFCD00000826
0.03% max.
01,130; 08,92; 15,84
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
HEDRZPFGACZZDS-UHFFFAOYSA-N
trichloromethane
6212
119.38
Spectroscopy

Chemical Identifiers

67-66-3
119.37
HEDRZPFGACZZDS-UHFFFAOYSA-N
6212
ClC(Cl)Cl
CHCl3
MFCD00000826
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
CHEBI:35255
Videos
Safety and Handling

Safety and Handling

GHS P Statement Causes skin irritation. Causes serious eye irritation. Harmful if swallowed. Toxic if inhaled. Suspected of causing cancer. May cause drowsiness or dizziness. Suspected of damaging the unborn child. Causes damage to organs through prolonged or repeated exposure if inhaled.

GHS P Statement IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician. IF ON SKIN: Wash with plenty of soap and water. Do not breathe dust/fume/gas/mist/vapors/spray. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection.

Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only