missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chloroform, 99+%, Thermo Scientific Chemicals

$145.00 - $1,406.35

Chemical Identifiers

CAS 67-66-3
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.37
MDL Number MFCD00000826
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
PubChem CID 6212
ChEBI CHEBI:35255
IUPAC Name chloroform
SMILES ClC(Cl)Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC158210010
SDS View Documents Promotion Details
Thermo Scientific Chemicals
158210010
1 L Glass Bottle N/A
 
AC158210025
SDS View Documents
Thermo Scientific Chemicals
158210025
2.5 L Glass Bottle N/A
 
AC158210250
SDS View Documents
Thermo Scientific Chemicals
158210250
25 L Plastic Can N/A
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

67-66-3
119.37
HEDRZPFGACZZDS-UHFFFAOYSA-N
6212
chloroform
CHCl3
MFCD00000826
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
CHEBI:35255
ClC(Cl)Cl

Specifications

-63.0°C
290°C
61.0°C
Glass Bottle
1.4443 to 1.4463
99+%
CHCl3
MFCD00000826
0.56 mPa.s (20°C)
01, 61
15, 2142
Solubility in water: 8g/L (20°C). Other solubilities: miscible with alcohol, benzene, ether, petr.ether,, carbon tetrachloride, carbon disulfide and oils., soluble in aceton
ClC(Cl)Cl
119.37
CHEBI:35255
≥99%
Chloroform
67-66-3,64-17-5
1.4800g/mL
1 L
Liquid
0.002% max.
1.48
CHCl3
Vapor Pressure: 213mbar at 20°C
0.02% max. (K.F.)
01,130; 08,92; 15,84
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
HEDRZPFGACZZDS-UHFFFAOYSA-N
chloroform
6212
119.38
Extra Pure
Videos
Safety and Handling

Safety and Handling

GHS P Statement Harmful if swallowed. Toxic if inhaled. Causes skin irritation. Causes serious eye irritation. Suspected of causing cancer. Suspected of damaging the unborn child. Causes damage to organs through prolonged or repeated exposure if inhaled.

GHS P Statement IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection.

Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only