missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chloroform, ACS Grade, ≥ 99.8%, LabChem™

Supplier:  LabChem LC130401

 View more versions of this product

Catalog No. LC130401


Explore available promotions

Description

Description

Specifications

Specifications

Chloroform
67-66-3,64-17-5
99,1
1,490kg/m3
500 mL
Amber Glass
≤0.001%
CHCl3
MFCD00000826
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
ClC(Cl)Cl
119.37
CHEBI:35255
≥99.8%
-64°C
Colorless
Chlorine
61°C
Passes Test
Liquid
1490kg/m3
CHCl3
UN1888
HEDRZPFGACZZDS-UHFFFAOYSA-N
trichloromethane
6212
119.38
ACS

Chemical Identifiers

67-66-3
119.37
HEDRZPFGACZZDS-UHFFFAOYSA-N
6212
trichloromethane
CHCl3
MFCD00000826
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
CHEBI:35255
ClC(Cl)Cl
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Toxic if inhaled.
Causes skin and serious eye irritation.
Suspected of causing cancer.
Suspected of damaging the unborn child.
Causes damage to organs (liver, kidneys) through prolonged or repeated exposure.

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Do not breathe vapors.
Do not eat, drink or smoke when using this product.
Use only outdoors or in a well-ventilated area.
Wear eye protection, face protection, protective clothing, protective gloves.
Wash exposed skin thoroughly after handling.
If exposed or concerned: Get medical advice/attention.
If swallowed: Rinse mouth.
Call a poison center or doctor if you feel unwell.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing and wash it before reuse.
If skin irritation occurs: Get medical attention.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center or doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation

Danger

EINECSNumber : 200-663-8

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions