Learn More
Chloroform (Ethanol as Preservative, Spectranalyzed™), Fisher Chemical™
$829.58 - $829.58
Chemical Identifiers
CAS | 67-66-3 |
---|---|
Molecular Formula | CHCl3 |
Molecular Weight (g/mol) | 119.37 |
MDL Number | MFCD00000826 |
InChI Key | HEDRZPFGACZZDS-UHFFFAOYSA-N |
Synonym | trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio |
PubChem CID | 6212 |
ChEBI | CHEBI:35255 |
IUPAC Name | trichloromethane |
SMILES | ClC(Cl)Cl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
C574-4
|
Fisher Chemical
BPC5744 |
4 L | Glass Bottle |
Each for $829.58
Case of 4 Each for $3,318.32
|
|
||||
Chemical Identifiers
67-66-3 | |
119.37 | |
HEDRZPFGACZZDS-UHFFFAOYSA-N | |
6212 | |
trichloromethane |
CHCl3 | |
MFCD00000826 | |
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio | |
CHEBI:35255 | |
ClC(Cl)Cl |
Specifications
-63°C | |
Pass Test | |
290°C | |
61°C | |
Pass Test | |
Glass Bottle | |
2ppm max. | |
CHCl3 | |
213mbar at 20°C | |
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio | |
ClC(Cl)Cl | |
119.37 | |
CHEBI:35255 | |
≥99.8% | |
Approx. 0.75% Ethanol as preservative |
67-66-3,64-17-5 | |
Colorless | |
1.480g/cm³ | |
4 L | |
Conforms to Ref | |
Liquid | |
≥99.8 % | |
MFCD00000826 | |
0.56 mPa/s at 20°C | |
HEDRZPFGACZZDS-UHFFFAOYSA-N | |
trichloromethane | |
6212 | |
119.38 | |
Also meets ACS specifications | |
Chloroform, Approx. 0.75% ethanol as preservative |
Safety and Handling
WARNING!
Emergency Overview
Possible cancer hazard. May cause cancer based on animal data. Irritating to eyes and skin. May cause central nervous system effects. Aspiration hazard if swallowed - can enter lungs and cause damage. May be harmful if inhaled. May cause irritation of respiratory tract. Danger of serious damage to health by prolonged exposure. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention.
NFPA
Health:2
Flammability:1
Instability:0
DOTInformation : DOT Class 6.1, : Poison