Learn More
Chloroform, HPLC Grade, 99.5+% min, stab. with amylene, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 043685K7
Description
Chloroform is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Chloroform | |
67-66-3 | |
1.492 | |
4 L | |
Characteristic | |
1.446 | |
CHCl3 | |
158mm Hg at 20°C | |
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio | |
HEDRZPFGACZZDS-UHFFFAOYSA-N | |
chloroform | |
6212 | |
119.38 |
°C to 63°C | |
Colorless | |
60.5°C to 61.5°C | |
982°C (1800°F) | |
Liquid | |
>99.5% | |
MFCD00000826 | |
1731042 | |
Miscible with diethyl ether,oils,ligroin,alcohol,carbon tetrachloride and carbon disulfide. | |
ClC(Cl)Cl | |
119.37 | |
CHEBI:35255 | |
HPLC |
Chemical Identifiers
67-66-3 | |
119.37 | |
HEDRZPFGACZZDS-UHFFFAOYSA-N | |
6212 | |
chloroform |
CHCl3 | |
MFCD00000826 | |
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio | |
CHEBI:35255 | |
ClC(Cl)Cl |
Safety and Handling
P201-P202-P260-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c
H302-H315-H319-H331-H335-H336-H351-H361d-H373
DOTInformation : DOT Class: 6.1, Packaging Group: III
EINECSNumber : 200-663-8
RTECSNumber : FS9100000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only