missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chloroform, Reagent ACS, 99.8+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 423555000

 View more versions of this product

Catalog No. AC423555000



Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Chloroform
Stabilized
290°C
61.0°C
Conforms to Ref Report
Liquid
≤0.001%
1.48
CHCl3
Vapor Pressure: 213mbar at 20°C
01, 61
15, 2142
Solubility in water: 8g/L (20°C). Other solubilities: miscible with alcohol, benzene, ether, petr.ether,, carbon tetrachloride, carbon disulfide and oils., soluble in aceton
ClC(Cl)Cl
119.37
CHEBI:35255
≥99.8%
-63.0°C
67-66-3,64-17-5
1.4800g/mL
500 mL
Glass Bottle
1.4443 to 1.4463
99.8+%
CHCl3
MFCD00000826
0.56 mPa.s (20°C)
01,130; 08,92; 15,84
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
HEDRZPFGACZZDS-UHFFFAOYSA-N
chloroform
6212
119.38
ACS Reagent

Chemical Identifiers

67-66-3
119.37
HEDRZPFGACZZDS-UHFFFAOYSA-N
6212
ClC(Cl)Cl
CHCl3
MFCD00000826
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
CHEBI:35255
Videos
Safety and Handling

Safety and Handling

GHS P Statement Harmful if swallowed. Toxic if inhaled. Causes skin irritation. Causes serious eye irritation. Suspected of causing cancer. Suspected of damaging the unborn child. Causes damage to organs through prolonged or repeated exposure if inhaled.

GHS P Statement IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection.

Warning

EINECSNumber : 200-663-8

RTECSNumber : FS9100000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only