Learn More
Chloroform, Spectrophotometric Grade, 99.5+%, Thermo Scientific Chemicals
$115.30 - $300.49
Chemical Identifiers
CAS | 67-66-3 |
---|---|
Molecular Formula | CHCl3 |
Molecular Weight (g/mol) | 119.37 |
MDL Number | MFCD00000826 |
InChI Key | HEDRZPFGACZZDS-UHFFFAOYSA-N |
Synonym | trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio |
PubChem CID | 6212 |
ChEBI | CHEBI:35255 |
IUPAC Name | chloroform |
SMILES | ClC(Cl)Cl |
Description
Chloroform is used in spectrophotometry and environmental testing.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
67-66-3 | |
119.37 | |
HEDRZPFGACZZDS-UHFFFAOYSA-N | |
6212 | |
chloroform |
CHCl3 | |
MFCD00000826 | |
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio | |
CHEBI:35255 | |
ClC(Cl)Cl |
Specifications
°C to 63°C | |
Colorless | |
1.492 g/cm3 at 20°C | |
1 L | |
Liquid | |
>99.5% | |
MFCD00000826 | |
1731042 | |
Miscible with diethyl ether,oils,ligroin,alcohol,carbon tetrachloride and carbon disulfide. | |
ClC(Cl)Cl | |
119.37 | |
CHEBI:35255 | |
Spectrophotometric |
67-66-3 | |
Colorless liquid | |
60.5°C to 61.5°C | |
Characteristic | |
1.446 | |
CHCl3 | |
210 hPa (158mm Hg) at 20°C | |
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio | |
HEDRZPFGACZZDS-UHFFFAOYSA-N | |
chloroform | |
6212 | |
119.38 | |
Chloroform |
Safety and Handling
Wear suitable protective clothing and gloves.
P201-P202-P260-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c
H302-H315-H319-H331-H335-H336-H351-H361d-H373
DOTInformation : DOT Class: 6.1; Packing Group: III
EINECSNumber : 200-663-8
RTECSNumber : FS9100000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only