missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chloromethanesulfonyl chloride, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 04148604

 View more versions of this product

Catalog No. AA4148604


Explore available promotions

Description

Description

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuff. It is synthesis of chloromethanesulfonates and chloromethanesulfonamides. It is also used for conversion of alcohols into the inverted acetates, azides and nitriles, and rearrangement of carbon-carbon and carbon-oxygen bonds under mild conditions, and in electrophilic and radical reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Chloromethanesulfonyl chloride
Colorless to Yellow
78°C to 81°C (20 mmHg)
Acidic
ClCH2SO2Cl
2 g
1811761
methanesulfonyl chloride, chloro, chloromethylsulfonyl chloride, chloromethanesulphonyl chloride, chlormethansulfochlorid, chlormethansulfonylchlorid, chloromesyl chloride, chloromethyl sulfonyl chloride, chlormethansulfochlorid czech, chlormethansulfonylchlorid czech, chlorid kyseliny chlormethansulfonove
KQDDQXNVESLJNO-UHFFFAOYSA-N
chloromethanesulfonyl chloride
77054
98%
3518-65-8
1.67 g/mL
98%
CH2Cl2O2S
MFCD00014727
UN3265
Moisture sensitive
It is soluble in dichloromethane,pyridine,and most organic solvents. It is insoluble in water.
ClCS(Cl)(=O)=O
148.99
149
Liquid

Chemical Identifiers

3518-65-8
148.99
KQDDQXNVESLJNO-UHFFFAOYSA-N
77054
ClCS(Cl)(=O)=O
CH2Cl2O2S
MFCD00014727
methanesulfonyl chloride, chloro, chloromethylsulfonyl chloride, chloromethanesulphonyl chloride, chlormethansulfochlorid, chlormethansulfonylchlorid, chloromesyl chloride, chloromethyl sulfonyl chloride, chlormethansulfochlorid czech, chlormethansulfonylchlorid czech, chlorid kyseliny chlormethansulfonove
chloromethanesulfonyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302+H312+H332-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 222-526-1

RTECSNumber : PB2800000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only