Learn More
(Chloromethyl)cyclopropane, 97%, Thermo Scientific Chemicals
$168.38 - $598.99
Chemical Identifiers
CAS | 5911-08-0 |
---|---|
Molecular Formula | C4H7Cl |
Molecular Weight (g/mol) | 90.55 |
MDL Number | MFCD00001307 |
InChI Key | ZVTQWXCKQTUVPY-UHFFFAOYSA-N |
Synonym | chloromethyl cyclopropane, cylopropylmethyl chloride, cyclopropylmethyl chloride, cyclopropane, chloromethyl, chloromethyl-cyclopropane, j7okqcc0kw, unii-j7okqcc0kw, cylopropylmethylchloride, chlorocyclopropylmethane, cyclopropylchloromethane |
PubChem CID | 80013 |
SMILES | ClCC1CC1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAL0698606
|
Thermo Scientific Chemicals
L0698606 |
5 g |
N/A
|
|
||||||
AAL0698614
|
Thermo Scientific Chemicals
L0698614 |
25 g |
N/A
|
|
||||||
Description
(Chloromethyl)cyclopropane was used in the synthesis of (lithiomethyl)cyclopropane and homoallyllithium via 4,4?-di-tert-butylbiphenyl or naphthalene-catalysed lithiation reaction. On reaction with isolable dialkylsilylene affords 5-silaspiro[4.4]nonane derivative.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(Chloromethyl)cyclopropane was used in the synthesis of (lithiomethyl)cyclopropane and homoallyllithium via 4,4′-di-tert-butylbiphenyl or naphthalene-catalysed lithiation reaction. On reaction with isolable dialkylsilylene affords 5-silaspiro[4.4]nonane derivative.
Solubility
Not miscible in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, bases and heat.
Chemical Identifiers
5911-08-0 | |
90.55 | |
ZVTQWXCKQTUVPY-UHFFFAOYSA-N | |
80013 |
C4H7Cl | |
MFCD00001307 | |
chloromethyl cyclopropane, cylopropylmethyl chloride, cyclopropylmethyl chloride, cyclopropane, chloromethyl, chloromethyl-cyclopropane, j7okqcc0kw, unii-j7okqcc0kw, cylopropylmethylchloride, chlorocyclopropylmethane, cyclopropylchloromethane | |
ClCC1CC1 |
Specifications
5911-08-0 | |
0.98 | |
−1°C (30°F) | |
C4H7Cl | |
MFCD00001307 | |
UN1993 | |
chloromethyl cyclopropane, cylopropylmethyl chloride, cyclopropylmethyl chloride, cyclopropane, chloromethyl, chloromethyl-cyclopropane, j7okqcc0kw, unii-j7okqcc0kw, cylopropylmethylchloride, chlorocyclopropylmethane, cyclopropylchloromethane | |
ZVTQWXCKQTUVPY-UHFFFAOYSA-N | |
chloromethylcyclopropane | |
80013 | |
97% |
-91°C | |
87°C to 89°C | |
Recognizable | |
1.436 | |
5 g | |
635697 | |
Not miscible in water. | |
ClCC1CC1 | |
90.55 | |
90.55 | |
(Chloromethyl)cyclopropane |
Safety and Handling
GHS H Statement
H225-H302-H315-H319-H335
Highly flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P330-P370+P378q-P501c
H225-H302
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 227-632-1
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only