missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(Chloromethyl)triethoxysilane 95.0+%, TCI America™
$41.24 - $118.56
Chemical Identifiers
CAS | 15267-95-5 |
---|---|
Molecular Formula | C7H17ClO3Si |
Molecular Weight (g/mol) | 212.75 |
MDL Number | MFCD00039354 |
InChI Key | ZDOBWJOCPDIBRZ-UHFFFAOYSA-N |
PubChem CID | 84858 |
IUPAC Name | (chloromethyl)triethoxysilane |
SMILES | CCO[Si](CCl)(OCC)OCC |
Chemical Identifiers
15267-95-5 | |
212.75 | |
ZDOBWJOCPDIBRZ-UHFFFAOYSA-N | |
(chloromethyl)triethoxysilane |
C7H17ClO3Si | |
MFCD00039354 | |
84858 | |
CCO[Si](CCl)(OCC)OCC |
Specifications
15267-95-5 | |
180°C | |
MFCD00039354 | |
1993 | |
CCO[Si](CCl)(OCC)OCC | |
212.75 | |
212.75 | |
Liquid |
Yellow | |
C7H17ClO3Si | |
10 g | |
ZDOBWJOCPDIBRZ-UHFFFAOYSA-N | |
(chloromethyl)triethoxysilane | |
84858 | |
≥95.0% (GC) | |
(Chloromethyl)triethoxysilane |
Safety and Handling
EINECSNumber : (2)-2079
RTECSNumber : VV2275000
TSCA : Yes