Learn More
Chloromethyltrimethylsilane, 98+%, Thermo Scientific Chemicals
$129.36 - $435.23
Chemical Identifiers
CAS | 2344-80-1 |
---|---|
Molecular Formula | C4H11ClSi |
Molecular Weight (g/mol) | 122.67 |
MDL Number | MFCD00000878 |
InChI Key | OOCUOKHIVGWCTJ-UHFFFAOYSA-N |
Synonym | chloromethyl trimethylsilane, silane, chloromethyl trimethyl, trimethylsilyl methyl chloride, chloromethyl trimethyl silane, ch3 3sich2cl, tmcms, pubchem13668, trimethylchloromethylsilane |
PubChem CID | 75361 |
IUPAC Name | chloromethyl(trimethyl)silane |
SMILES | C[Si](C)(C)CCl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC194760250
|
Thermo Scientific Chemicals
194760250 |
25 g |
Each for $129.36
|
|
|||||
AC194761000
|
Thermo Scientific Chemicals
194761000 |
100 g |
Each for $435.23
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagentChemical Identifiers
2344-80-1 | |
122.67 | |
OOCUOKHIVGWCTJ-UHFFFAOYSA-N | |
75361 | |
C[Si](C)(C)CCl |
C4H11ClSi | |
MFCD00000878 | |
chloromethyl trimethylsilane, silane, chloromethyl trimethyl, trimethylsilyl methyl chloride, chloromethyl trimethyl silane, ch3 3sich2cl, tmcms, pubchem13668, trimethylchloromethylsilane | |
chloromethyl(trimethyl)silane |
Specifications
2344-80-1 | |
0.8790g/mL | |
−2°C | |
98+% | |
C4H11ClSi | |
(CH3)3SiCH2Cl | |
25 g | |
12,118; 13,76 | |
chloromethyl trimethylsilane, silane, chloromethyl trimethyl, trimethylsilyl methyl chloride, chloromethyl trimethyl silane, ch3 3sich2cl, tmcms, pubchem13668, trimethylchloromethylsilane | |
OOCUOKHIVGWCTJ-UHFFFAOYSA-N | |
chloromethyl(trimethyl)silane | |
75361 | |
98+% | |
Chloromethyltrimethylsilane, 98+% |
Colorless | |
97.0°C to 98.0°C | |
Authentic | |
Glass bottle | |
1.4165 to 1.4185 | |
MFCD00000878 | |
04, III, 1844 | |
0.879 | |
Solubility in water: insoluble | |
C[Si](C)(C)CCl | |
122.67 | |
122.67 | |
Liquid |
Safety and Handling
GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 219-058-5
TSCA : TSCA
RUO – Research Use Only