missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chlorophyllin, coppered trisodium salt, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1615506

 View more versions of this product

Catalog No. AAA1615506

Explore available promotions

Description

Description

Chlorophyllin, coppered trisodium salt is used in the spectrophotometric determination of Ven- lafaxine hydrochloride. A hybrid photoelectrochemical biofuel cell is composed of a TiO 2 film conductive glass electrode sensitized by a copper chlorophyll trisodium salt. Visible light-sensitive gels were prepared by introducing a chromophore (trisodium salt of copper chlorophyllin) to thermosensitive gels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorophyllin, coppered trisodium salt is used in the spectrophotometric determination of Ven- lafaxine hydrochloride. A hybrid photoelectrochemical biofuel cell is composed of a TiO 2 film conductive glass electrode sensitized by a copper chlorophyll trisodium salt. Visible light-sensitive gels were prepared by introducing a chromophore (trisodium salt of copper chlorophyllin) to thermosensitive gels.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.
Specifications

Specifications

Chlorophyllin
11006-34-1
C34H31CuN4Na3O6
5 g
724.15
coppered trisodium salt
Odorless
MFCD00012149
Soluble in water.

Chemical Identifiers

11006-34-1
C34H31CuN4Na3O6
MFCD00012149
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 234-242-5

RTECSNumber : GS2168866

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only