Learn More
Chlorophyllin, coppered trisodium salt
CAS: 11006-34-1 | C34H31CuN4Na3O6
Supplier: Thermo Scientific Chemicals A1615522
Description
Chlorophyllin, coppered trisodium salt is used in the spectrophotometric determination of Ven- lafaxine hydrochloride. A hybrid photoelectrochemical biofuel cell is composed of a TiO 2 film conductive glass electrode sensitized by a copper chlorophyll trisodium salt. Visible light-sensitive gels were prepared by introducing a chromophore (trisodium salt of copper chlorophyllin) to thermosensitive gels.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsChlorophyllin, coppered trisodium salt is used in the spectrophotometric determination of Ven- lafaxine hydrochloride. A hybrid photoelectrochemical biofuel cell is composed of a TiO 2 film conductive glass electrode sensitized by a copper chlorophyll trisodium salt. Visible light-sensitive gels were prepared by introducing a chromophore (trisodium salt of copper chlorophyllin) to thermosensitive gels.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.
Specifications
| Chlorophyllin | |
| 11006-34-1 | |
| C34H31CuN4Na3O6 | |
| 100 g | |
| 724.15 |
| coppered trisodium salt | |
| Odorless | |
| MFCD00012149 | |
| Soluble in water. |
Chemical Identifiers
| 11006-34-1 | |
| C34H31CuN4Na3O6 | |
| MFCD00012149 |
Safety and Handling
EINECSNumber : 234-242-5
RTECSNumber : GS2168866
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only

