missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chloroplatinic Acid Hexahydrate (Certified ACS), Fisher Chemical

Supplier:  Fisher Chemical P1541

Catalog No. P154-1



Explore available promotions

Description

Description

Specifications

Specifications

Chloroplatinic Acid Hexahydrate
18497-13-7,16941-12-1,26023-84-7
Powder/Solid
Cl6H14O6Pt
UN 2507
Pass Test (in ethanol)
[H+].[H+].O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Pt+4]
517.89
≥37.50%
Pass Test
2.43g/mL
60°C
Amber
1 g
MFCD00149910
Dihydrogen Hexachloroplatinum (IV) Hexahydrate
PIJUVEPNGATXOD-UHFFFAOYSA-J
platinum(4+) dihydrogen hexahydrate hexachloride
517.91
Certified ACS
Glass Ampoule
Videos
Safety and Handling

Safety and Handling

Emergency Overview
Toxic if swallowed. Causes burns by all exposure routes. May cause allergic respiratory and skin reaction. May cause sensitization by inhalation and skin contact. Water reactive. Contact with water liberates toxic gas.
NFPA
Health:3 Flammability:0 Instability:1

Signal Word: DANGER!

DOTInformation : DOT Class 8, : Corrosive

SDS Documents

Documents

Promotions

Promotions