missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chloroquine diphosphate salt, 98%, Thermo Scientific Chemicals

Chloroquine diphosphate salt, CAS # 50-63-5, is a member of the quinolone family that is considered to be a DNA intercalator. It shows anti-malarial, anti-inflammatory, and anti-filaricides activities.

$213.66 - $1048.23

Chemical Identifiers

CAS 50-63-5
Molecular Formula C18H32ClN3O8P2
Molecular Weight (g/mol) 515.86
MDL Number MFCD00069852
InChI Key QKICWELGRMTQCR-UHFFFAOYNA-N
Synonym chloroquine diphosphate, chloroquine phosphate, aralen phosphate, chingamin phosphate, chloroquine bis phosphate, chingaminum, alermine, h-stadur, aralen diphosphate, miniquine
PubChem CID 64927
SMILES OP(O)(O)=O.OP(O)(O)=O.CCN(CC)CCCC(C)NC1=C2C=CC(Cl)=CC2=NC=C1
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6445914
View Documents
Thermo Scientific Chemicals
J6445914
25 g
Each for $213.66
 
AAJ6445918
View Documents
Thermo Scientific Chemicals
J6445918
50 g
Each for $321.41
 
AAJ6445922
View Documents
Thermo Scientific Chemicals
J6445922
100 g
Each for $505.85
 
AAJ6445930
View Documents
Thermo Scientific Chemicals
J6445930
250 g
Each for $1,048.23
 
Description

Description

Chloroquine diphosphate salt is used to study the role of endosomal acidification in cellular processes, such as the signaling of intracellular TLRs. It can be used as DNA intercalator & to dissociate antigen antibody complexes without denaturing red blood cell antigens. Chloroquine showed very high antiviral activity against NiV but very little activity against the other viruses at concentrations lower than 20 μM.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description
• Chloroquine diphosphate salt is a DNA intercalator that shows anti-malarial and anti-inflammatory activities.
• It has been described that this compound interferes with the biosynthesis of nucleic acids, suggesting a cellular mechanism of action
Applications
• Chloroquine diphosphate salt is used as a DNA intercalator to dissociate antigen-antibody complexes without denaturing red blood cell antigens
• This compound shows antimalarial activity against Plasmodium falciparum, except against chloroquine-resistant strains. It has been used in in vitro antiplasmodial assays. In parasites, the mechanism of action remains unclear, but this compound can inhibit the parasitic enzyme heme polymerase that converts the toxic heme into non-toxic hemozoin, resulting in the accumulation of toxic heme within the parasite.
• It shows very high anti-viral activity against NiV but lower activity against other viruses at concentrations lower than 20 μM
• Chloroquine diphosphate salt is used to study the role of endosomal acidification in cellular processes, such as the signaling of intracellular TLRs
• Chloroquine diphosphate salt has been used in autophagy inhibition studies
Specifications

Chemical Identifiers

50-63-5
515.86
QKICWELGRMTQCR-UHFFFAOYNA-N
64927
C18H32ClN3O8P2
MFCD00069852
chloroquine diphosphate, chloroquine phosphate, aralen phosphate, chingamin phosphate, chloroquine bis phosphate, chingaminum, alermine, h-stadur, aralen diphosphate, miniquine
OP(O)(O)=O.OP(O)(O)=O.CCN(CC)CCCC(C)NC1=C2C=CC(Cl)=CC2=NC=C1

Specifications

50-63-5
White
MFCD00069852
4223142
chloroquine diphosphate, chloroquine phosphate, aralen phosphate, chingamin phosphate, chloroquine bis phosphate, chingaminum, alermine, h-stadur, aralen diphosphate, miniquine
QKICWELGRMTQCR-UHFFFAOYNA-N
4-N-(7-chloroquinolin-4-yl)-1-N,1-N-diethylpentane-1,4-diamine;phosphoric acid
64927
98%
Chloroquine diphosphate salt
213°C to 216°C
C18H32ClN3O8P2
25 g
14,2163
Soluble in water
OP(O)(O)=O.OP(O)(O)=O.CCN(CC)CCCC(C)NC1=C2C=CC(Cl)=CC2=NC=C1
515.86
515.87
Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

P264b-P270-P301+P312-P330-P501c

H302

EINECSNumber : 200-055-2

RTECSNumber : VB2450000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only