missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Berkshire™ Choice™ Cleanroom Socks

Choice™ Cleanroom Socks are made of continuous filament polyester yarn and are laundered and packaged in a Class 5 cleanroom.

Supplier:  Berkshire™ CHSOCKR1050

Catalog No. 19-266-710


Explore available promotions

Description

Description

  • 99% Continuous filament polyester, 1% spandex
  • One size fits all
  • Mid-calf length with elastic cuff for secure fit
  • Soft and comfortable
  • Laundered and packaged in an ISO Class 5 cleanroom
  • Color: White
  • 10 pairs per pack

Technology

  • Constructed with continuous filament yarns to reduce fiber and particle shedding
  • Cleanroom laundered and packaged

Advantages

  • Meets the requirements of EU GMP Annex 1 section 7.14
  • Cleanroom laundry process reduces fiber and particle shedding and minimizes bioburden
  • Over 1000 times cleaner than regular polyester/cotton socks
  • Comfortable and durable for all day wear
  • A cost effective, clean and disposable alternative to operators own socks
  • Autoclavable (121°C, 30 minutes)
  • Bulk packaging available
Specifications

Specifications

99% Continuous filament polyester, 1% spandex
One Size
Videos
SDS
Documents

Documents

Promotions

Promotions