Thermo Scientific Chemicals Cholecystokinin, CCK Octapeptide (26-33)
For cellular and molecular biology applications
Supplier:  Thermo Scientific Chemicals J66669EXD
 View more versions of this product
Catalog No. AAJ66669EXD
Explore available promotions
Description
Description
Cholecystokinin, CCK Octapeptide (26-33) is used in the biological studies in the preparation of opioid receptors agonistic and CCK receptor antagonistic activity. It plays a role in satiety.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
HPLC
-30°C to -10°C
1143.27
0.5 mg
Asp-Tyr(SO3H)-Met-Gly-Trp-Met-Asp-Phe-NH2
CCK 8
Solid
Cholecystokinin, CCK Octapeptide (26-33)
RUO - research use only