missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cholesterol, NF Grade, MP Biomedicals
$61.44 - $61.44
Chemical Identifiers
| CAS | 57-88-5 |
|---|---|
| Molecular Formula | C27H46O |
| Molecular Weight (g/mol) | 386.664 |
| MDL Number | MFCD00003646 |
| InChI Key | HVYWMOMLDIMFJA-DPAQBDIFSA-N |
| Synonym | cholesterol, cholesterin, cholest-5-en-3beta-ol, cholesteryl alcohol, cholestrin, cordulan, dusoline, dusoran, provitamin d, cholesterine |
| PubChem CID | 5997 |
| ChEBI | CHEBI:16113 |
| IUPAC Name | (3S,8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-ol |
| SMILES | CC(C)CCCC(C)C1CCC2C1(CCC3C2CC=C4C3(CCC(C4)O)C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
MP21013803
|
MP Biomedicals Inc
021013803 |
25 g |
Each for $61.44
|
|
|||||
Description
- Cholesterol is suitable for use in liposome preparations
- Cholesterol is a component used for the preparation and study of artificial model membranes
- One example is a study of valinomycin induced changes in membrane potentials of red blood cell and phospholipid (phosphatidylcholine from egg yolk plus cholesterol) vesicle suspensions
- Positively charged cyanine dyes were used that fluorimetrically respond to the change in potential
Chemical Identifiers
| 57-88-5 | |
| 386.664 | |
| HVYWMOMLDIMFJA-DPAQBDIFSA-N | |
| 5997 | |
| (3S,8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-ol |
| C27H46O | |
| MFCD00003646 | |
| cholesterol, cholesterin, cholest-5-en-3beta-ol, cholesteryl alcohol, cholestrin, cordulan, dusoline, dusoran, provitamin d, cholesterine | |
| CHEBI:16113 | |
| CC(C)CCCC(C)C1CCC2C1(CCC3C2CC=C4C3(CCC(C4)O)C)C |
Specifications
| 57-88-5 | |
| White | |
| C27H46O | |
| MFCD00003646 | |
| cholesterol, cholesterin, cholest-5-en-3beta-ol, cholesteryl alcohol, cholestrin, cordulan, dusoline, dusoran, provitamin d, cholesterine | |
| CC(C)CCCC(C)C1CCC2C1(CCC3C2CC=C4C3(CCC(C4)O)C)C | |
| 386.664 | |
| CHEBI:16113 | |
| NF | |
| CHOLESTEROL |
| 147°C to 150°C | |
| 360°C (literature) | |
| C27H46O | |
| 25 g | |
| HVYWMOMLDIMFJA-DPAQBDIFSA-N | |
| (3S,8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-ol | |
| 5997 | |
| 386.67 | |
| Powder |
Safety and Handling
Recommended Storage : -20°C.