missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cholesteryl nonanoate, Thermo Scientific Chemicals

$93.27 - $264.79

Chemical Identifiers

CAS 1182-66-7
Molecular Formula C36H62O2
Molecular Weight (g/mol) 526.89
MDL Number MFCD00003643
InChI Key WCLNGBQPTVENHV-MKQVXYPISA-N
Synonym cholesteryl pelargonate, cholesteryl nonanoate, cholesterol pelargonate, unii-4313o7p4xw, 5-cholesten-3beta-ol 3-nonanoate, cholesterol, nonanoate, 3beta-hydroxy-5-cholestene 3-nonanoate, nonanoic acid cholesterol ester, pelargonic acid cholesterol ester, cholest-5-ene-3-beta-yl nonanoate
PubChem CID 2723614
IUPAC Name [(3S,8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-yl] nonanoate
SMILES CCCCCCCCC(=O)OC1CCC2(C3CCC4(C(C3CC=C2C1)CCC4C(C)CCCC(C)C)C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0285714
View Documents
Thermo Scientific Chemicals
L0285714
25 g
Each for $93.27
 
AAL0285722
View Documents
Thermo Scientific Chemicals
L0285722
100 g
Each for $264.79
 
Description

Description

Cholesteryl nonanoate is used in hair colors, cosmetic products like lip products, lotions. It is also used in the manufature of some pleochroic dyes. It can be also used as a component of the liquid crystals used for liquid crystal displays.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cholesteryl nonanoate is used in hair colors, cosmetic products like lip products, lotions. It is also used in the manufature of some pleochroic dyes. It can be also used as a component of the liquid crystals used for liquid crystal displays.

Solubility
Soluble in Chloroform, Tetrahydrofuran(THF). Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents, Strong acids, Strong bases, Strong reducing agents.
Specifications

Chemical Identifiers

1182-66-7
526.89
WCLNGBQPTVENHV-MKQVXYPISA-N
2723614
CCCCCCCCC(=O)OC1CCC2(C3CCC4(C(C3CC=C2C1)CCC4C(C)CCCC(C)C)C)C
C36H62O2
MFCD00003643
cholesteryl pelargonate, cholesteryl nonanoate, cholesterol pelargonate, unii-4313o7p4xw, 5-cholesten-3beta-ol 3-nonanoate, cholesterol, nonanoate, 3beta-hydroxy-5-cholestene 3-nonanoate, nonanoic acid cholesterol ester, pelargonic acid cholesterol ester, cholest-5-ene-3-beta-yl nonanoate
[(3S,8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-yl] nonanoate

Specifications

1182-66-7
Odorless
MFCD00003643
3179820
Soluble in Chloroform,Tetrahydrofuran(THF). Insoluble in water.
−30° (c=5 in Chloroform)
[(3S,8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-yl] nonanoate
2723614
Cholesteryl nonanoate
75°C to 79°C
C36H62O2
25 g
cholesteryl pelargonate, cholesteryl nonanoate, cholesterol pelargonate, unii-4313o7p4xw, 5-cholesten-3beta-ol 3-nonanoate, cholesterol, nonanoate, 3beta-hydroxy-5-cholestene 3-nonanoate, nonanoic acid cholesterol ester, pelargonic acid cholesterol ester, cholest-5-ene-3-beta-yl nonanoate
WCLNGBQPTVENHV-MKQVXYPISA-N
CCCCCCCCC(=O)OC1CCC2(C3CCC4(C(C3CC=C2C1)CCC4C(C)CCCC(C)C)C)C
526.89
526.89
Safety and Handling

Safety and Handling

EINECSNumber : 214-658-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only