missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Choline bitatrate, >98%, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0210138490

 View more versions of this product

Catalog No. ICN10138490


Explore available promotions

Description

Description

Specifications

Specifications

Choline bitatrate
87-67-2
>98%
MFCD00036332
choline bitartrate, cholini bitartras, choline 3-carboxy-2,3-dihydroxypropanoate, 2-hydroxyethyl trimethylammonium bitartrate, choline tartrate 1:1, 2-hydroxy-n,n,n-trimethylethanaminium 3-carboxy-2,3-dihydroxypropanoate, cholinibitatratis, choline hydrogen l-+-tartrate, ethanaminium, 2-hydroxy-n,n,n-trimethyl-, 2r,3r-2,3-dihydroxybutanedioate 1:1
C[N+](C)(C)CCO.C(C(C(=O)[O-])O)(C(=O)O)O
253.251
253.3
Molecular biology; Cell biology
White
C9H19NO7
500 g
QWJSAWXRUVVRLH-UHFFFAOYSA-M
2-hydroxyethyl(trimethyl)azanium;2,3,4-trihydroxy-4-oxobutanoate
10198924
Powder

Chemical Identifiers

87-67-2
253.251
QWJSAWXRUVVRLH-UHFFFAOYSA-M
10198924
C[N+](C)(C)CCO.C(C(C(=O)[O-])O)(C(=O)O)O
C9H19NO7
MFCD00036332
choline bitartrate, cholini bitartras, choline 3-carboxy-2,3-dihydroxypropanoate, 2-hydroxyethyl trimethylammonium bitartrate, choline tartrate 1:1, 2-hydroxy-n,n,n-trimethylethanaminium 3-carboxy-2,3-dihydroxypropanoate, cholinibitatratis, choline hydrogen l-+-tartrate, ethanaminium, 2-hydroxy-n,n,n-trimethyl-, 2r,3r-2,3-dihydroxybutanedioate 1:1
2-hydroxyethyl(trimethyl)azanium;2,3,4-trihydroxy-4-oxobutanoate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 201-763-4

Recommended Storage : Store at room temperature (15° to 30°C), desiccate.

SDS
Documents

Documents

Promotions

Promotions