missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific Chemicals Choline hydroxide, 45 wt.% aqueous solution, pure, stabilized

$155.21 - $155.21

Specifications

Chemical Name or Material Choline hydroxide, 45 wt.% aqueous solution
CAS 123-41-1,7732-18-5
Color White
Density 1.0700g/mL
Molecular Formula C5H15NO2
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC386721000
View Documents Promotion Details
Thermo Scientific Chemicals
386721000
100 mL Glass bottle
Each for $155.21
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Choline hydroxide, 45 wt.% aqueous solution
White
C5H15NO2
15,2211
choline hydroxide, gossypine, sincaline, bursine, luridine, sinkalin, sinkaline, fagine, vidine, 2-hydroxy-n,n,n-trimethylethanaminium hydroxide
KIZQNNOULOCVDM-UHFFFAOYSA-M
2-hydroxyethyl(trimethyl)azanium;hydroxide
1800ppm ethylenediamine 3000ppm hydroxylamine
121.18
Pure
123-41-1,7732-18-5
1.0700g/mL
MFCD00002831
1.07
Solubility in water: miscible.
[OH-].C[N+](C)(C)CCO
121.18
31255
44.0 to 48.0% (Total base)
Granular Powder
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only