missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

ChromaCare™ LC-MS Biologics Flush Solution, Thermo Scientific™

Flush solution for cleaning blood, plasma, and urine samples from the chromatograph.

$997.37 - $1022.33

Specifications

CAS 67-64-1,75-05-8
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
Percent Purity Pass Test (By GC-FID)
Grade LC-MS
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
MB1241
View Documents
Thermo Fisher Scientific Formerly Fairlawn
MB1241
1 L Borosilicate Glass Bottle
Each for $1,022.33
Case of 6 Each for $6,133.98
 
MB124212
View Documents
Thermo Fisher Scientific Formerly Fairlawn
MB124212
2.5 L Amber Glass Bottle
Each for $997.37
Case of 4 Each for $3,989.48
 
Description

Description

  • Suitable for flushing difficult to clean samples such as blood, plasma, and urine after vitamin D, hormone or drugs of abuse testing
  • Save time by using off-the-shelf blend — premade for convenience
  • Helps to reduce sample carryover
  • Packaged in 2.5L amber glass bottle
Specifications

Specifications

67-64-1,75-05-8
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
Pass Test (By GC-FID)
LC-MS
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only