missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chroman-6-carboxylic acid, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC32801CB

 View more versions of this product

Catalog No. CC32801CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

Chroman-6-carboxylic acid
96.5
97%
C10H10O3
250mg
IFKANGOXGBPILW-UHFFFAOYSA-N
3,4-dihydro-2H-chromene-6-carboxylic acid
6504216
97%
103203-84-5
100.0
Amber Glass Bottle
MFCD08146590
chroman-6-carboxylic acid, 3,4-dihydro-2h-1-benzopyran-6-carboxylic acid, chromane-6-carboxylic acid, chroman-6-carboxylicacid, 2h-1-benzopyran-6-carboxylicacid, 3,4-dihydro, 6-chromanecarboxylic acid, acmc-1c8ke, 3,4-dihydro-2h-benzopyran-6-carboxylic acid
OC(=O)C1=CC2=C(OCCC2)C=C1
178.19
178.19

Chemical Identifiers

103203-84-5
178.19
IFKANGOXGBPILW-UHFFFAOYSA-N
6504216
OC(=O)C1=CC2=C(OCCC2)C=C1
C10H10O3
MFCD08146590
chroman-6-carboxylic acid, 3,4-dihydro-2h-1-benzopyran-6-carboxylic acid, chromane-6-carboxylic acid, chroman-6-carboxylicacid, 2h-1-benzopyran-6-carboxylicacid, 3,4-dihydro, 6-chromanecarboxylic acid, acmc-1c8ke, 3,4-dihydro-2h-benzopyran-6-carboxylic acid
3,4-dihydro-2H-chromene-6-carboxylic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS Documents

Documents

Promotions

Promotions