Ahlstrom-Munksjo Chromatography, Electrophoresis and Blotting Paper, Grade 238
Medium thick paper is recommended for general chromatography application. Also ideal for use as wicks in protein or nucleic acid blotting to provide a uniform flow or buffer through the gel to transfer membrane in a blotting sandwich.
Supplier: Ahlstrom-Munksjo 23882020
Specifications
Chromatography applications. Use as wicks in protein or nucleic acid blotting. Suited for electrophoresis | |
0.35 mm | |
200 mm |
238 | |
200 x 200 mm | |
200 mm |