missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromic-Sulfuric Acid Cleaning Solution for Cleaning Laboratory Glassware, Reagents
Chromic-Sulfuric Acid Cleaning Solution for Cleaning Laboratory Glassware, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc CC0048001C1
Specifications
| No | |
| 0 | |
| Not Autoclavable | |
| Brown | |
| 1 L | |
| 1 L |
| Liquid | |
| Non-sterile | |
| 260°C | |
| Safety Coated Glass | |
| Completely miscible |