missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chromium(II) chloride, 90%, Thermo Scientific Chemicals

$348.54 - $348.54

Chemical Identifiers

CAS 10049-05-5
Molecular Formula Cl2Cr
Molecular Weight (g/mol) 122.9
MDL Number MFCD00010947
Synonym Chromous chloride
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC223810100
SDS View Documents
Thermo Scientific Chemicals
223810100
10 g Glass bottle
Each for $348.54
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
Specifications

Chemical Identifiers

10049-05-5
122.9
Chromous chloride
Cl2Cr
MFCD00010947

Specifications

824.0°C
Gray to Green
10 g
Cl2Cr
MFCD00010947
15, 2248
Solubility in water: may decompose. Other solubilities: insoluble in alcohol and ether
122.9
Glass bottle
10049-05-5
Powder
88% min. (Argentometry)
CrCl2
01,149; 02,76; 03,60; 04,144; 07,73; 09,511; 12,136; 13,84; 14,94; 16,93; 17,84
Chromous chloride
122.9
90%
Chromium(II) chloride, 90%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
If eye irritation persists: Get medical

GHS Signal Word: Warning

EINECSNumber : 233-163-3

RTECSNumber : GB5250000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only