Learn More
Chromium(II) chloride, anhydrous, 97%, Thermo Scientific Chemicals
$146.95 - $1393.30
Chemical Identifiers
CAS | 10049-05-5 |
---|---|
Molecular Formula | CrCl2 |
MDL Number | MFCD00010947 |
Synonym | Chromous chloride |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1434206
|
Thermo Scientific Chemicals
L1434206 |
5 g |
Each for $146.95
|
|
|||||
AAL1434214
|
Thermo Scientific Chemicals
L1434214 |
25 g |
Each for $515.14
|
|
|||||
AAL1434222
|
Thermo Scientific Chemicals
L1434222 |
100 g |
Each for $1,393.30
|
|
|||||
Description
Chromium(II) chloride is used as a precursor to other inorganic and organometallic chromium complexes. It is a reagent in the Nozaki-Hiyama-Kishi reaction. It is also used in the Takai olefination to prepare vinyl iodides from aldehydes in the presence of iodoform.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsChromium(II) chloride is used as a precursor to other inorganic and organometallic chromium complexes. It is a reagent in the Nozaki-Hiyama-Kishi reaction. It is also used in the Takai olefination to prepare vinyl iodides from aldehydes in the presence of iodoform.
Solubility
Soluble in water. Insoluble in alcohol and ether.
Notes
Hygroscopic. Incompatible with strong oxidizing agents, water and active metals.
Chemical Identifiers
10049-05-5 | |
CrCl2 | |
MFCD00010947 | |
Chromous chloride |
Specifications
824°C | |
5 g | |
MFCD00010947 | |
14,2244 | |
Soluble in water. Insoluble in alcohol and ether. | |
97% | |
Air and moisture sensitive | |
Chromium(II) chloride, anhydrous |
10049-05-5 | |
CrCl2 | |
UN3260 | |
Chromous chloride | |
122.9 | |
Odorless | |
2.878 |
Safety and Handling
GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber : 233-163-3
RTECSNumber : GB5250000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only