Learn More
Chromium(III) chloride hexahydrate, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01068606
Description
Chromium(III) chloride hexahydrate is used as an organic synthesis catalyst and as an additive to intravenous solutions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Chromium(III) chloride hexahydrate | |
10060-12-5 | |
Crystalline Aggregates | |
(metals basis) | |
CrCl3·6H2O | |
UN3260 | |
chromium iii chloride hexahydrate, chromic chloride hexahydrate, hexaaquachromium chloride, chromium trichloride hexahydrate, trichlorochromium hexahydrate, chromium chloride hexahydrate, hexaaquachromium iii chloride, chrom iii-chlorid, chromium 3+ chloride hexahydrate, chlorid chromity hexahydrat czech | |
LJAOOBNHPFKCDR-UHFFFAOYSA-K | |
chromium(3+) hexahydrate trichloride | |
104957 | |
266.45 (158.35 Anhydrous) | |
Puratronic™ | |
1.835 g/mL |
83°C | |
Green | |
5 g | |
Cl3CrH12O6 | |
MFCD00149660 | |
14,2222 | |
Soluble in water and alcohol. Insoluble in diethyl ether. | |
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cr+3] | |
266.44 | |
CHEBI:53442 | |
99.998% | |
Odorless |
Chemical Identifiers
10060-12-5 | |
266.44 | |
LJAOOBNHPFKCDR-UHFFFAOYSA-K | |
104957 | |
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cr+3] |
Cl3CrH12O6 | |
MFCD00149660 | |
chromium iii chloride hexahydrate, chromic chloride hexahydrate, hexaaquachromium chloride, chromium trichloride hexahydrate, trichlorochromium hexahydrate, chromium chloride hexahydrate, hexaaquachromium iii chloride, chrom iii-chlorid, chromium 3+ chloride hexahydrate, chlorid chromity hexahydrat czech | |
CHEBI:53442 |
Safety and Handling
GHS H Statement
H334-H314-H318-H302-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
P234-P261-P264b-P270-P272-P280g-P301+P312-P302+P352-P330-P333+P313-P363-P390-P501c
H290-H302-H317
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber : 233-038-3
RTECSNumber : GB5450000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only