Learn More
Chromium(III) chloride tetrahydrofuran complex (1:3), 98%, Thermo Scientific™
Chemical Identifiers
| CAS | 10170-68-0 |
|---|---|
| Molecular Formula | C12H24Cl3CrO3 |
| Molecular Weight (g/mol) | 374.67 |
| MDL Number | MFCD00013222 |
| InChI Key | CYOMBOLDXZUMBU-UHFFFAOYSA-K |
| Synonym | chromium iii chloride tetrahydrofuran complex 1:3, chromium iii chloride tetrahydrofuran adduct, crcl3 thf 3, crcl3 3thf, chromium iii chloride tetrahydrofuran complex, chromium chloride; tris tetrahydrofuran, chromium iii chloride complex 1:3 tetrahydrofuran |
| PubChem CID | 11100829 |
| IUPAC Name | oxolane;trichlorochromium |
| SMILES | C1CCOC1.C1CCOC1.C1CCOC1.Cl[Cr](Cl)Cl |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC389850100
|
Thermo Scientific Chemicals
389850100 |
10g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 10170-68-0 | |
| 374.67 | |
| CYOMBOLDXZUMBU-UHFFFAOYSA-K | |
| 11100829 | |
| C1CCOC1.C1CCOC1.C1CCOC1.Cl[Cr](Cl)Cl |
| C12H24Cl3CrO3 | |
| MFCD00013222 | |
| chromium iii chloride tetrahydrofuran complex 1:3, chromium iii chloride tetrahydrofuran adduct, crcl3 thf 3, crcl3 3thf, chromium iii chloride tetrahydrofuran complex, chromium chloride; tris tetrahydrofuran, chromium iii chloride complex 1:3 tetrahydrofuran | |
| oxolane;trichlorochromium |
Specifications
| 10170-68-0 | |
| 98% | |
| MFCD00013222 | |
| CYOMBOLDXZUMBU-UHFFFAOYSA-K | |
| oxolane;trichlorochromium | |
| 11100829 | |
| 98% | |
| Glass bottle |
| 10g | |
| C12H24Cl3CrO3 | |
| chromium iii chloride tetrahydrofuran complex 1:3, chromium iii chloride tetrahydrofuran adduct, crcl3 thf 3, crcl3 3thf, chromium iii chloride tetrahydrofuran complex, chromium chloride; tris tetrahydrofuran, chromium iii chloride complex 1:3 tetrahydrofuran | |
| C1CCOC1.C1CCOC1.C1CCOC1.Cl[Cr](Cl)Cl | |
| 374.67 | |
| 374.67 | |
| Authentic | |
| Chromium(III) chloride tetrahydrofuran complex (1:3) |
Safety and Handling
GHS H Statement
Flammable solid.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
May form explosive peroxides.
GHS P Statement
Eliminate all ignition sources if safe to do so.
Take precautionary measures against static discharge.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protec
GHS Signal Word: Warning
RUO â Research Use Only