missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chromium(III) fluoride, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 03574104

 View more versions of this product

Catalog No. AA3574104


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Chromium(III) fluoride
Green
Powder
(metals basis)
UN1756
chromium fluoride, chromic fluoride, chromium 3+ trifluoride, acmc-1bht2, chromium 3+ ion trifluoride, chromic fluoride, solid un1756 corrosive, chromic fluoride, solution un1757 corrosive
99.98%
Hygroscopic
>1000°C
7788-97-8
2 g
MFCD00016045
14,2223
108.99
Odorless
3.8 g/mL

Chemical Identifiers

7788-97-8
MFCD00016045
chromium fluoride, chromic fluoride, chromium 3+ trifluoride, acmc-1bht2, chromium 3+ ion trifluoride, chromic fluoride, solid un1756 corrosive, chromic fluoride, solution un1757 corrosive
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302+H312+H332-H314

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHROMIC FLUORIDE, SOLID

EINECSNumber : 232-137-9

RTECSNumber : GB6125000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only