Learn More
Chromium(III) nitrate nonahydrate, 98.5%, Thermo Scientific Chemicals
For Use As a Catalyst, Corrosion Inhibitor, in Textile Printing
$75.70 - $316.32
Chemical Identifiers
CAS | 7789-02-8 |
---|---|
Molecular Formula | CrN3O9 |
Molecular Weight (g/mol) | 238.01 |
MDL Number | MFCD00149659 |
InChI Key | PHFQLYPOURZARY-UHFFFAOYSA-N |
Synonym | chromium iii nitrate nonahydrate, unii-d2806iol1l, nitric acid, chromium 3+ salt, nonahydrate, acmc-20alg5, bis 7-chlorophenyl sulfone, cr.3no3.9h2o, ksc495a9j, chromium trinitrate, nonahydrate, chromium iii nitrate, nonahydrate, chromium 3+ nonahydrate trinitronate |
PubChem CID | 9887206 |
IUPAC Name | chromium(3+);trinitrate;nonahydrate |
SMILES | [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Cr+3] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA1156514
|
Thermo Scientific Chemicals
01156514 |
25 g |
Each for $75.70
|
|
|||||
AA1156530
|
Thermo Scientific Chemicals
01156530 |
250 g |
Each for $105.84
|
|
|||||
AA11565A1
|
Thermo Scientific Chemicals
011565A1 |
1 kg |
Each for $316.32
|
|
|||||
Description
Chromium nitrate is used in the manufacture of alkali metal-free catalysts and in preservation. It is used in the preparation of chrome catalysts, in textile printing operations, and as a corrosion inhibitor. Chromium(III) nitrate as a source of chromium is added to food supplements for nutritional purposes .
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7789-02-8 | |
238.01 | |
PHFQLYPOURZARY-UHFFFAOYSA-N | |
9887206 | |
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Cr+3] |
CrN3O9 | |
MFCD00149659 | |
chromium iii nitrate nonahydrate, unii-d2806iol1l, nitric acid, chromium 3+ salt, nonahydrate, acmc-20alg5, bis 7-chlorophenyl sulfone, cr.3no3.9h2o, ksc495a9j, chromium trinitrate, nonahydrate, chromium iii nitrate, nonahydrate, chromium 3+ nonahydrate trinitronate | |
chromium(3+);trinitrate;nonahydrate |
Specifications
60°C | |
Purple | |
25 g | |
CrN3O9 | |
chromium iii nitrate nonahydrate, unii-d2806iol1l, nitric acid, chromium 3+ salt, nonahydrate, acmc-20alg5, bis 7-chlorophenyl sulfone, cr.3no3.9h2o, ksc495a9j, chromium trinitrate, nonahydrate, chromium iii nitrate, nonahydrate, chromium 3+ nonahydrate trinitronate | |
PHFQLYPOURZARY-UHFFFAOYSA-N | |
chromium(3+);trinitrate;nonahydrate | |
9887206 | |
Odorless | |
Chromium(III) nitrate nonahydrate, nonahydrate |
7789-02-8 | |
Crystalline | |
98.5% | |
MFCD00149659 | |
Soluble in water and alcohol. | |
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Cr+3] | |
238.01 | |
400.14 (238.01 Anhydrous) | |
Purple crystalline |
Safety and Handling
P210-P220-P221-P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P363-P370+P378q-P501c
H272-H317-H319-H332
DOTInformation : DOT Class: 5.1, Packaging Group: III
EINECSNumber : 236-921-1
RTECSNumber : GB6300000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only