missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chromium(III) nitrate nonahydrate, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 219201000

 View more versions of this product

Catalog No. AC219201000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Chromium(III) nitrate nonahydrate
99%
Black to Purple
Crystalline Chunks
98.5% min. (Iodometry)
Cr(NO3)3·9H2O
14,2226
Solubility in water: 1g/L (20°C). Other solubilities: soluble in alcohol
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Cr+3]
400.14
400.14
Plastic bottle
60.0°C
7789-02-8
2 to 3 (5% aq. soln.)
100 g
CrN3O9·9H2O
MFCD00149659
chromium iii nitrate nonahydrate, unii-d2806iol1l, nitric acid, chromium 3+ salt, nonahydrate, acmc-20alg5, bis 7-chlorophenyl sulfone, cr.3no3.9h2o, ksc495a9j, chromium trinitrate, nonahydrate, chromium iii nitrate, nonahydrate, chromium 3+ nonahydrate trinitronate
PHFQLYPOURZARY-UHFFFAOYSA-N
chromium(3+);trinitrate;nonahydrate
9887206
99%

Chemical Identifiers

7789-02-8
PHFQLYPOURZARY-UHFFFAOYSA-N
9887206
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Cr+3]
MFCD00149659
chromium iii nitrate nonahydrate, unii-d2806iol1l, nitric acid, chromium 3+ salt, nonahydrate, acmc-20alg5, bis 7-chlorophenyl sulfone, cr.3no3.9h2o, ksc495a9j, chromium trinitrate, nonahydrate, chromium iii nitrate, nonahydrate, chromium 3+ nonahydrate trinitronate
chromium(3+);trinitrate;nonahydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if inhaled.
Causes serious eye irritation.
May intensify fire; oxidizer.
May cause an allergic skin reaction.
Toxic to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Take any precaution to avoid mixing with combustibles.
Call a POISON CE

GHS Signal Word: Warning

RTECSNumber : GB6300000

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only