missing translation for 'onlineSavingsMsg'
Learn More

Chromium powder, APS <10 micron, 99.2% (metals basis)

CAS: 7440-47-3 | Cr | 52.00 g/mol

$112.45 - $1071.53

Chemical Identifiers

CAS 7440-47-3
Molecular Formula Cr
Molecular Weight (g/mol) 52.00
MDL Number MFCD00010944
InChI Key VYZAMTAEIAYCRO-UHFFFAOYSA-N
Synonym chrom, chrome, cromo, metal, chrome french, compounds, chrom german, chromium, elemental, chromium, metal, ccris 159
PubChem CID 23976
ChEBI CHEBI:28073
IUPAC Name chromium
SMILES [Cr]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Qty  
AA4179718
View Documents
Thermo Scientific Chemicals
04179718
50 g
Each for $112.45
Only null left
 
AA4179730
View Documents
Thermo Scientific Chemicals
04179730
250 g
Each for $365.42
Only null left
 
AA41797A1
View Documents
Thermo Scientific Chemicals
041797A1
1 kg
Each for $1,071.53
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-47-3
52.00
VYZAMTAEIAYCRO-UHFFFAOYSA-N
23976
chromium
Cr
MFCD00010944
chrom, chrome, cromo, metal, chrome french, compounds, chrom german, chromium, elemental, chromium, metal, ccris 159
CHEBI:28073
[Cr]

Specifications

Gray
Gray powder
Odorless
99.2% (Metals basis)
MFCD00010944
VYZAMTAEIAYCRO-UHFFFAOYSA-N
chromium
23976
Chromium
7440-47-3
50 g
Powder
Cr
chrom, chrome, cromo, metal, chrome french, compounds, chrom german, chromium, elemental, chromium, metal, ccris 159
[Cr]
52.00
CHEBI:28073
Videos
Safety and Handling

Safety and Handling

Wear suitable protective clothing and gloves.

P210-P240-P241-P280-P370+P378q

H228

DOTInformation : DOT Class: 4.1, Packaging Group: III

EINECSNumber : 231-157-5

RTECSNumber : GB4200000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only