missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hexavalent Chromium ICP Standard, 10,000 ppm Cr6+ in 5% HCl, Ricca Chemical

Supplier:  Ricca Chemical Company PCR610KH50

 View more versions of this product

Catalog No. PCR610KH-50

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

Chromium(VI) ICP Standard
Natural Poly Bottle
90.0,5.34,2.19
Orange-Yellow
JOSWYUNQBRPBDN-UHFFFAOYSA-P
diazanium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
24600
7789-09-5,7647-01-0,7789-09-5
50 mL
94.0,5.56,2.28
Cr2H8N2O7
[NH4+].[NH4+].[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-]
252.06
1mL = 10mg Cr (10, 000ppm Cr)(NH4)2Cr2O7 in 5% HCl

Chemical Identifiers

7789-09-5
252.06
24600
[NH4+].[NH4+].[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-]
Cr2H8N2O7
JOSWYUNQBRPBDN-UHFFFAOYSA-P
diazanium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
Videos
Safety and Handling

Safety and Handling

ShelfLife : 18 months

SDS
Documents

Documents

Promotions

Promotions