Learn More
Chromium(VI) oxide, 99%, Thermo Scientific Chemicals
$68.22 - $445.97
Chemical Identifiers
CAS | 1333-82-0 |
---|---|
Molecular Formula | CrO3 |
Molecular Weight (g/mol) | 99.99 |
MDL Number | MFCD00010952 |
InChI Key | WGLPBDUCMAPZCE-UHFFFAOYSA-N |
Synonym | chromium trioxide, chromium vi oxide, monochromium trioxide, chromtrioxid, chromic trioxide, chromsaeureanhydrid, anhydride chromique, chromium 6+ oxide, chromium vi trioxide, chromium oxide |
PubChem CID | 14915 |
ChEBI | CHEBI:48240 |
IUPAC Name | trioxochromium |
SMILES | O=[Cr](=O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA1252230
|
Thermo Scientific Chemicals
01252230 |
250 g |
Each for $68.22
|
|
|||||
AA12522A1
|
Thermo Scientific Chemicals
012522A1 |
1 kg |
Each for $133.63
|
|
|||||
AA12522D9
|
Thermo Scientific Chemicals
012522D9 |
5 x 1 kg |
Each for $445.97
|
|
|||||
Description
It is widely used in chromium electroplating, and as protective coatings in corrosion/oxidation resistance of metals. It is also employed in CDs and DVDs. It is useful as an oxidant in Jones oxidation in acetic acid or acetone. It is used in the oxidation of primary alcohols into carboxylic acids, and secondary alcohols into ketones. With phosphoric acid, it is used as a stripping agent for anodic coatings of all types and in the production of syntheric rubies.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1333-82-0 | |
99.99 | |
WGLPBDUCMAPZCE-UHFFFAOYSA-N | |
14915 | |
trioxochromium |
CrO3 | |
MFCD00010952 | |
chromium trioxide, chromium vi oxide, monochromium trioxide, chromtrioxid, chromic trioxide, chromsaeureanhydrid, anhydride chromique, chromium 6+ oxide, chromium vi trioxide, chromium oxide | |
CHEBI:48240 | |
O=[Cr](=O)=O |
Specifications
250°C (decomposition) | |
1333-82-0 | |
250 g | |
CrO3 | |
chromium trioxide, chromium vi oxide, monochromium trioxide, chromtrioxid, chromic trioxide, chromsaeureanhydrid, anhydride chromique, chromium 6+ oxide, chromium vi trioxide, chromium oxide | |
WGLPBDUCMAPZCE-UHFFFAOYSA-N | |
trioxochromium | |
14915 | |
99.99 | |
Dark red crystals | |
Chromium(VI) oxide |
196°C | |
Crystalline | |
99% | |
MFCD00010952 | |
Very soluble in water. Soluble in sulfuric,nitric acid,acetic acid,and acetone. | |
O=[Cr](=O)=O | |
99.99 | |
CHEBI:48240 | |
Hygroscopic | |
2.7 g/cm3 |
Safety and Handling
P201-P202-P210-P220-P221-P260-P262-P264b-P270-P271-P272-P280g-P281-P283-P284-P285-P302+P350-P304+P340-P305+P351+P338-P306+P360-P310-P330-P331-P333+P313-P353-P363-P370+P378q-P3
H271-H301-H310+H330-H314-H317-H334-H335-H340-H350-H361f-H372
DOTInformation : DOT Class: 5.1, Packaging Group: II
EINECSNumber : 215-607-8
RTECSNumber : GB6650000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only