missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Chrysin 98.0+%, TCI America™
Supplier: TCI America C165225G
Specifications
Chrysin | |
C15H10O4 | |
25 g | |
RTIXKCRFFJGDFG-UHFFFAOYSA-N | |
5,7-dihydroxy-2-phenylchromen-4-one | |
5281607 | |
254.24 | |
Crystalline Powder |
480-40-0 | |
MFCD00006834 | |
chrysin, 5,7-dihydroxyflavone, chrysine, 5,7-dihydroxy-2-phenyl-4h-chromen-4-one, crysin, 4h-1-benzopyran-4-one, 5,7-dihydroxy-2-phenyl, unii-3cn01f5zj5, flavone, 5,7-dihydroxy, 5,7-dihydroxy-2-phenyl-4h-1-benzopyran-4-one, chembl117 | |
C1=CC=C(C=C1)C2=CC(=O)C3=C(C=C(C=C3O2)O)O | |
254.241 | |
CHEBI:75095 | |
≥98.0% (HPLC,T) |
Chemical Identifiers
480-40-0 | |
254.241 | |
RTIXKCRFFJGDFG-UHFFFAOYSA-N | |
5281607 | |
5,7-dihydroxy-2-phenylchromen-4-one |
C15H10O4 | |
MFCD00006834 | |
chrysin, 5,7-dihydroxyflavone, chrysine, 5,7-dihydroxy-2-phenyl-4h-chromen-4-one, crysin, 4h-1-benzopyran-4-one, 5,7-dihydroxy-2-phenyl, unii-3cn01f5zj5, flavone, 5,7-dihydroxy, 5,7-dihydroxy-2-phenyl-4h-1-benzopyran-4-one, chembl117 | |
CHEBI:75095 | |
C1=CC=C(C=C1)C2=CC(=O)C3=C(C=C(C=C3O2)O)O |
Safety and Handling
RTECSNumber : LK8329050
TSCA : No