Velp™ Scientific Chuck Guard for OHS 20 Digital Overhead Stirrer
Protection guard that prevents the operator from contacting the stirring elements.
Supplier:  Velp™ Scientific A00001312
Catalog No. 11392187
Explore available promotions
Description
Description
  • Protects operators from contact with upper part of chuck
  • Prevents potential entanglement with rotating shafts
  • Easy-to-use: just fix the support plate using two screws
  • Robust construction
  • Light weight
Specifications
Specifications
OHS 20 Digital Overhead Stirrer
0.095 kg
Plexiglass
Chuck Guard
69 x 75 x 120 mm
0.21 lb.