Thermo Scientific™ Cimarec+™ Hotplate Series
Get exceptional safety and temperature performance for your routine protocols.
$578.21 - $1838.15
Specifications
Display LED
Warranty 3 Years
Type Hot Plate
Frequency 50/60 Hz
View More Specs
Products 4
Catalog Number Mfr. No. Size Top Plate Material Plug Type Price Quantity  
Catalog Number Mfr. No. Size Top Plate Material Plug Type Price Quantity  
HP88854100
View Documents Promotion Details
Thermo Scientific™
HP88854100
4.25 x 4.25 in. Ceramic US-Style
Each for $578.21
HP88857100
View Documents Promotion Details
Thermo Scientific™
HP88857100
7.25 x 7.25 in. Ceramic US-Style
Each for $628.78
HP88857104
View Documents
Thermo Scientific™
HP88857104
7.25 x 7.25 in. Aluminum US-Style
Each for $813.60
HP88850100
View Documents Promotion Details
Thermo Scientific™
HP88850100
10.25 x 10.25 in. Ceramic US-Style
Each for $1,838.15
Description
Description
Get exceptional safety and temperature performance for your routine protocols with the Thermo Scientific™ Cimarec+™ Hotplate series. You'll find exceptional safety and temperature performance for your routine protocols. The Cimarec+ Series are available in three sizes, providing flexibility from microscale chemistries to production operations.
Advanced technology
  • Temperature adjustable in 1 degree increments
  • Hot Top warning system protects from accidental burns with prominent display when heating surface is above 50°C (122°F)
Flexible, robust design
  • Top plate choices in ceramic or aluminum
  • Easy-to-read LED display for heating
  • Raised display design protects electronics from spills
  • Optional temperature probe and splash guard protection shield
  • Variable sizes for different sample volumes
Warranty: 3 years
Specifications
Specifications
LED
3 Years
Hot Plate
50/60 Hz
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.