Cincinnati Surgical Swann Morton Sterile Stainless Steel Blades and Scalpels
For use in wide variety of medical applications, including Dermaplaning
Supplier: Cincinnati Surgical 01SM11
Description
For use in wide variety of medical applications, including Dermaplaning
- Blades packaged with VCI Paper to protect blade from corrosion
- Scalpels individually packaged in transparent thermos formed tray
- 200% visual inspection
- Strongest/Smallest foil package available
- Narrow grinding angle resulting in increased initial keenness
- High surface finish
- Cellophane overwrap
- Extended range
Specifications
Stainless Steel | |
# 11 | |
Packaged with VCI Paper to protect Blade from Corrosion |
Blade | |
Sterile |