Cincinnati Surgical Swann Morton Sterile Stainless Steel Blades and Scalpels
For use in wide variety of medical applications, including Dermaplaning
Supplier:  Cincinnati Surgical 01SM15
 View more versions of this product
Catalog No. 19200240
Explore available promotions
Description
Description
For use in wide variety of medical applications, including Dermaplaning
  • Blades packaged with VCI Paper to protect blade from corrosion
  • Scalpels individually packaged in transparent thermos formed tray
  • 200% visual inspection
  • Strongest/Smallest foil package available
  • Narrow grinding angle resulting in increased initial keenness
  • High surface finish
  • Cellophane overwrap
  • Extended range
Specifications
Specifications
Stainless Steel
# 15
Packaged with VCI Paper to protect Blade from Corrosion
Blade
Sterile