missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cinnamyl Acetate 97.0+%, TCI America™

$55.30 - $343.65

Chemical Identifiers

CAS 103-54-8
Molecular Formula C11H12O2
Molecular Weight (g/mol) 176.215
MDL Number MFCD00008722
InChI Key WJSDHUCWMSHDCR-VMPITWQZSA-N
Synonym cinnamyl acetate, 3-phenyl-2-propenyl acetate, 3-phenylallyl acetate, cinnamyl alcohol, acetate, acetic acid, cinnamyl ester, trans-cinnamyl acetate, 3-phenyl-2-propen-1-yl acetate, gamma-phenylallyl acetate, 1-acetoxy-3-phenyl-2-propene, 3-phenyl-2-propen-1-ol acetate
PubChem CID 5282110
IUPAC Name [(E)-3-phenylprop-2-enyl] acetate
SMILES CC(=O)OCC=CC1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A090125ML
View Documents
TCI America
A090125ML
25 mL N/A
 
A0901500ML
View Documents
TCI America
A0901500ML
500 mL N/A
 
Description

Description

Specifications

Chemical Identifiers

103-54-8
176.215
WJSDHUCWMSHDCR-VMPITWQZSA-N
5282110
CC(=O)OCC=CC1=CC=CC=C1
C11H12O2
MFCD00008722
cinnamyl acetate, 3-phenyl-2-propenyl acetate, 3-phenylallyl acetate, cinnamyl alcohol, acetate, acetic acid, cinnamyl ester, trans-cinnamyl acetate, 3-phenyl-2-propen-1-yl acetate, gamma-phenylallyl acetate, 1-acetoxy-3-phenyl-2-propene, 3-phenyl-2-propen-1-ol acetate
[(E)-3-phenylprop-2-enyl] acetate

Specifications

103-54-8
265°C
MFCD00008722
cinnamyl acetate, 3-phenyl-2-propenyl acetate, 3-phenylallyl acetate, cinnamyl alcohol, acetate, acetic acid, cinnamyl ester, trans-cinnamyl acetate, 3-phenyl-2-propen-1-yl acetate, gamma-phenylallyl acetate, 1-acetoxy-3-phenyl-2-propene, 3-phenyl-2-propen-1-ol acetate
CC(=O)OCC=CC1=CC=CC=C1
176.215
176.22
Liquid
Yellow
C11H12O2
25 mL
WJSDHUCWMSHDCR-VMPITWQZSA-N
[(E)-3-phenylprop-2-enyl] acetate
5282110
≥97.0% (GC)
Cinnamyl Acetate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1043

RTECSNumber : GE2275000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'