missing translation for 'onlineSavingsMsg'
Learn More

Cinnamyl alcohol, 98% trans

CAS: 104-54-1 | C9H10O | 134.18 g/mol

$57.58 - $57.58

Identifiants chimiques

CAS 104-54-1
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.18
MDL Number MFCD00002921
InChI Key OOCCDEMITAIZTP-QPJJXVBHSA-N
Synonym cinnamyl alcohol, cinnamic alcohol, 3-phenyl-2-propen-1-ol, 3-phenylprop-2-en-1-ol, zimtalcohol, styryl carbinol, e-3-phenylprop-2-en-1-ol, 3-phenylallyl alcohol, trans-cinnamyl alcohol, styryl alcohol
PubChem CID 5315892
ChEBI CHEBI:33227
IUPAC Name (E)-3-phenylprop-2-en-1-ol
SMILES C1=CC=C(C=C1)C=CCO
voir les spécifications

Produits 1
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Prix Quantité  
AC148521000
FDS Afficher les documents
Thermo Scientific Chemicals
148521000
100 g Glass bottle
chaque for $57.58
Il en reste null
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Identifiants chimiques

104-54-1
134.18
OOCCDEMITAIZTP-QPJJXVBHSA-N
5315892
(E)-3-phenylprop-2-en-1-ol
C9H10O
MFCD00002921
cinnamyl alcohol, cinnamic alcohol, 3-phenyl-2-propen-1-ol, 3-phenylprop-2-en-1-ol, zimtalcohol, styryl carbinol, e-3-phenylprop-2-en-1-ol, 3-phenylallyl alcohol, trans-cinnamyl alcohol, styryl alcohol
CHEBI:33227
C1=CC=C(C=C1)C=CCO

Spécifications

104-54-1
White
258.0°C
Authentic
C9H10O
MFCD00002921
06, 570
15, 2303
Solubility in water: 1.8g/L (20°C). Other solubilities: soluble in glycerol, methanol, clearly soluble in 3 volumes 50% alcohol, freely soluble in alcohol, ether, and other, common organic solvents
C1=CC=C(C=C1)C=CCO
134.18
CHEBI:33227
98% trans
Cinnamyl alcohol, 98%
31.0°C to 35.0°C
1.0400g/mL
126°C
Glass bottle
C6H5CH=CHCH2OH
100 g
1.04
cinnamyl alcohol, cinnamic alcohol, 3-phenyl-2-propen-1-ol, 3-phenylprop-2-en-1-ol, zimtalcohol, styryl carbinol, e-3-phenylprop-2-en-1-ol, 3-phenylallyl alcohol, trans-cinnamyl alcohol, styryl alcohol
OOCCDEMITAIZTP-QPJJXVBHSA-N
(E)-3-phenylprop-2-en-1-ol
5315892
134.18
Fused Low Melting Crystalline Solid
Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
May cause an allergic skin reaction.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Contaminated work clothing should not be allowed out of the workplace.
Avoid breathing dust/fume/gas/mist/vapors/spray.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 203-212-3

missing translation for 'rtecsNumber' : GE2200000

missing translation for 'tsca' : TSCA

FDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only