Learn More
Cinnamyl chloride, 95%, stab. with sodium carbonate, Thermo Scientific Chemicals
$100.99 - $1407.99
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1440818
|
Thermo Scientific Chemicals
A1440818 |
50 g |
Each for $100.99
|
|
|||||
AAA1440830
|
Thermo Scientific Chemicals
A1440830 |
250 g |
Each for $394.45
|
|
|||||
AAA144080B
|
Thermo Scientific Chemicals
A144080B |
1000 g |
Each for $1,407.99
|
|
|||||
Description
Cinnamyl chloride is used in the enantioselective total synthesis of helioporins C and E, bioactive marine diterpenes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCinnamyl chloride is used in the enantioselective total synthesis of helioporins C and E, bioactive marine diterpenes.
Solubility
Soluble in water (0.2 g/L at 20°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature: 2 - 8°C
Chemical Identifiers
2687-12-9 |
Specifications
2687-12-9 | |
1.096 | |
79°C (174°F) | |
1.5845 | |
50 g | |
741974 | |
Soluble in water (0.2g/L at 20°C). | |
C1=CC=C(C=C1)C=CCCl | |
152.62 | |
152.62 | |
Cinnamyl chloride, stabilized with sodium carbonate |
7°C | |
86°C to 87°C (2 mmHg) | |
C9H9Cl | |
MFCD00000986 | |
UN2927 | |
cinnamyl chloride, 3-chloroprop-1-en-1-yl benzene, 3-chloro-1-phenyl-1-propene, trans-cinnamyl chloride, 1e-3-chloroprop-1-en-1-yl benzene, benzene, 3-chloropropenyl, e-3-chloroprop-1-enyl benzene, unii-2i1adl56tx, 2i1adl56tx, cinnamylchloride | |
IWTYTFSSTWXZFU-QPJJXVBHSA-N | |
[(E)-3-chloroprop-1-enyl]benzene | |
639658 | |
95% |
Safety and Handling
GHS H Statement
H330-H314-H318-H227
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P235-P260-P264b-P270-P271-P272-P280-P284-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P501c
H227-H302-H314-H317-H330-H334-H335
DOTInformation : Hazard Class: 8; Packaging Group: I
EINECSNumber : 220-246-4
RTECSNumber : CZ0919905
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only