missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ciprofloxacin Hydrochloride Monohydrate 98.0+%, TCI America™
$94.56 - $701.76
Chemical Identifiers
CAS | 86393-32-0 |
---|---|
Molecular Formula | C17H21ClFN3O4 |
Molecular Weight (g/mol) | 385.82 |
MDL Number | MFCD00242856 |
InChI Key | ARPUHYJMCVWYCZ-UHFFFAOYSA-N |
Synonym | ciprofloxacin hydrochloride monohydrate, ciloxan, ceprimax, oftacilox, proquin xr, cipro, floxacipron, belmacina, ciflosin, ciprocinal |
PubChem CID | 62998 |
ChEBI | CHEBI:59936 |
IUPAC Name | hydrogen 1-cyclopropyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid hydrate chloride |
SMILES | [H+].O.[Cl-].OC(=O)C1=CN(C2CC2)C2=CC(N3CCNCC3)=C(F)C=C2C1=O |
Chemical Identifiers
86393-32-0 | |
385.82 | |
ARPUHYJMCVWYCZ-UHFFFAOYSA-N | |
62998 | |
hydrogen 1-cyclopropyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid hydrate chloride |
C17H21ClFN3O4 | |
MFCD00242856 | |
ciprofloxacin hydrochloride monohydrate, ciloxan, ceprimax, oftacilox, proquin xr, cipro, floxacipron, belmacina, ciflosin, ciprocinal | |
CHEBI:59936 | |
[H+].O.[Cl-].OC(=O)C1=CN(C2CC2)C2=CC(N3CCNCC3)=C(F)C=C2C1=O |
Specifications
86393-32-0 | |
C17H21ClFN3O4 | |
5 g | |
ARPUHYJMCVWYCZ-UHFFFAOYSA-N | |
hydrogen 1-cyclopropyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid hydrate chloride | |
62998 | |
367.81 | |
Crystalline Powder |
White-Yellow | |
MFCD00242856 | |
ciprofloxacin hydrochloride monohydrate, ciloxan, ceprimax, oftacilox, proquin xr, cipro, floxacipron, belmacina, ciflosin, ciprocinal | |
[H+].O.[Cl-].OC(=O)C1=CN(C2CC2)C2=CC(N3CCNCC3)=C(F)C=C2C1=O | |
385.82 | |
CHEBI:59936 | |
≥98.0% (HPLC,T) | |
Ciprofloxacin Hydrochloride Monohydrate |
Safety and Handling
TSCA : No