Fisherbrand™ Circular Cover Glasses
Made of the finest optical borosilicate glass, with uniform thickness and size.
$108.29 - $2012.00
Specifications
Shape | Circle |
---|---|
Material | Borosilicate Glass |
For Use With (Application) | Microscopy, Histology, Cytology, Hematology, Microbiology |
Quantity | 1 oz./Pk. |
Catalog Number | Mfr. No. | Thickness (Metric) | Diameter (Metric) | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Thickness (Metric) | Diameter (Metric) | Price | Quantity | |||||
12541000CA
|
Fisherbrand™
12CIR1602811T |
0.13 to 0.17 mm (#1) | 12 mm |
Pack for $321.89
Case of 10 Pack for $3,218.90
|
|
|||||
12541001CA
|
Fisherbrand™
12CIR15602811T |
0.16 to 0.19 mm (#1.5) | 12 mm |
Pack for $321.89
Case of 10 Pack for $3,218.90
|
|
|||||
12541002CA
|
Fisherbrand™
12CIR2602811T |
0.19 to 0.23 mm (#2) | 12 mm |
Pack of 10 Each for $153.23
Case of 10 Pack for $1,532.30
|
|
|||||
12541004CA
|
Fisherbrand™
15CIR2602811T |
0.19 to 0.23 mm (#2) | 15 mm |
Pack for $173.46
Case of 10 Pack for $1,734.60
|
|
|||||
12541005CA
|
Fisherbrand™
18CIR1602811T |
0.13 to 0.17 mm (#1) | 18 mm |
Pack for $164.34
Case of 10 Pack for $1,643.40
|
|
|||||
12541006CA
|
Fisherbrand™
18CIR2602811T |
0.19 to 0.23 mm (#2) | 18 mm |
Pack for $140.23
Case of 10 Pack for $1,402.30
|
|
|||||
12541008CA
|
Fisherbrand™
22CIR2602811T |
0.19 to 0.23 mm (#2) | 22 mm |
Pack for $108.29
Case of 10 Pack for $1,082.90
|
|
|||||
12541010CA
|
Fisherbrand™
25CIR2602811T |
0.19 to 0.23 mm (#2) | 25 mm |
Pack for $123.49
Case of 10 Pack for $1,234.90
|
|
|||||
12541003CA
|
Fisherbrand™
15CIR1602811T |
0.13 to 0.17 mm (#1) | 15 mm |
Case of 10 Each for $2,012.00
|
|
|||||
Description
All Circular Cover Slips feature a high-spectral transmission quality and finely adaptive refractive index suited for microscopy. Sold by weight, comes in 1 oz. packs.
- Manufactured with colorless borosilicate glass
- Available in a variety of sizes and thicknesses
Specifications
Circle | |
Borosilicate Glass | |
Microscopy, Histology, Cytology, Hematology, Microbiology | |
1 oz./Pk. |