missing translation for 'onlineSavingsMsg'
Learn More

cis-1,2-Cyclopentanediol, 98%, Thermo Scientific™

$91.14 - $299.14

Chemical Identifiers

CAS 5057-98-7
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.13
MDL Number MFCD00134192
InChI Key VCVOSERVUCJNPR-SYDPRGILSA-N
Synonym cis-1,2-cyclopentanediol, cis-cyclopentane-1,2-diol, cis-1,2-dihydroxycyclopentane, 1r,2s-cyclopentane-1,2-diol, 1s,2r cyclopentane-1,2-diol, cis-1,2-cyclopentandiol, 1,2-cis-cyclopentanediol, cis-1,2-cyclopen-tanediol, cis-cyclopenten-1,2-diol, 1alpha,2alpha-cyclopentanediol
PubChem CID 2733293
IUPAC Name (1R,2S)-cyclopentane-1,2-diol
SMILES C1CC(C(C1)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC310290010
SDS View Documents
Thermo Scientific Chemicals
310290010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC310290050
SDS View Documents
Thermo Scientific Chemicals
310290050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

5057-98-7
102.13
VCVOSERVUCJNPR-SYDPRGILSA-N
2733293
C1CC(C(C1)O)O
C5H10O2
MFCD00134192
cis-1,2-cyclopentanediol, cis-cyclopentane-1,2-diol, cis-1,2-dihydroxycyclopentane, 1r,2s-cyclopentane-1,2-diol, 1s,2r cyclopentane-1,2-diol, cis-1,2-cyclopentandiol, 1,2-cis-cyclopentanediol, cis-1,2-cyclopen-tanediol, cis-cyclopenten-1,2-diol, 1alpha,2alpha-cyclopentanediol
(1R,2S)-cyclopentane-1,2-diol

Specifications

5057-98-7
>110°C
97.5% min. (GC)
C5H10O2
1.4770 to 1.4790
1g
Solubility in water: partially soluble in water.
C1CC(C(C1)O)O
102.13
102.13
cis-1,2-Cyclopentanediol
83.0°C to 83.5°C (5.0mmHg)
Authentic
Glass bottle
C5H8(OH)2
MFCD00134192
cis-1,2-cyclopentanediol, cis-cyclopentane-1,2-diol, cis-1,2-dihydroxycyclopentane, 1r,2s-cyclopentane-1,2-diol, 1s,2r cyclopentane-1,2-diol, cis-1,2-cyclopentandiol, 1,2-cis-cyclopentanediol, cis-1,2-cyclopen-tanediol, cis-cyclopenten-1,2-diol, 1alpha,2alpha-cyclopentanediol
VCVOSERVUCJNPR-SYDPRGILSA-N
(1R,2S)-cyclopentane-1,2-diol
2733293
98%
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only