missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
cis-15-Tetracosenoic Acid 95.0+%, TCI America™
Supplier: TCI America T16421G
Specifications
cis-15-Tetracosenoic Acid | |
44°C | |
MFCD00010507 | |
nervonic acid, cis-15-tetracosenoic acid, z-tetracos-15-enoic acid, selacholeic acid, cis-selacholeic acid, 15z-tetracosenoic acid, unii-91oqs788be, z-15-tetracosenoic acid, 15-tetracosenoic acid | |
CCCCCCCCC=CCCCCCCCCCCCCCC(=O)O | |
366.63 | |
CHEBI:44247 | |
≥95.0% (GC) |
506-37-6 | |
C24H46O2 | |
1 g | |
GWHCXVQVJPWHRF-KTKRTIGZSA-N | |
(Z)-tetracos-15-enoic acid | |
5281120 | |
366.63 | |
Crystalline Powder |
Chemical Identifiers
506-37-6 | |
366.63 | |
GWHCXVQVJPWHRF-KTKRTIGZSA-N | |
5281120 | |
(Z)-tetracos-15-enoic acid |
C24H46O2 | |
MFCD00010507 | |
nervonic acid, cis-15-tetracosenoic acid, z-tetracos-15-enoic acid, selacholeic acid, cis-selacholeic acid, 15z-tetracosenoic acid, unii-91oqs788be, z-15-tetracosenoic acid, 15-tetracosenoic acid | |
CHEBI:44247 | |
CCCCCCCCC=CCCCCCCCCCCCCCC(=O)O |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator